Learn More
Tetraethylammonium Hydroxide, 10% in Water, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 420285000
48.26 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : 29
Additional Details : CAS Number : 77-98-5 Weight : 0.50000kg Transport : UN number : 2922 Chem class : 8 Pack group : II
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalystSpecifications
Tetraethylammonium hydroxide | |
10% in Water | |
102.0°C | |
(C2H5)4NOH | |
1.0200g/mL | |
tetraethylammonium hydroxide, tetraethyl ammonium hydroxide, unii-ra8vu41b1f, ammonium, tetraethyl-, hydroxide, tetraethylazanium hydroxide, ethanaminium, n,n,n-triethyl-, hydroxide, ra8vu41b1f, ethanaminium, n,n,n-triethyl-, hydroxide 1:1, tetraethylammonium hydroxide solution in water, n,n,n-triethylethanaminium | |
LRGJRHZIDJQFCL-UHFFFAOYSA-M | |
tetraethylazanium;hydroxide | |
6509 | |
9.5 to 10.5% | |
1.402 | |
500 g |
Colorless | |
77-98-5,7732-18-5 | |
C8H21NO | |
MFCD00009024 | |
15, 9346 | |
Solubility in water: soluble. | |
[OH-].CC[N+](CC)(CC)CC | |
147.26 | |
147.26 | |
Plastic bottle | |
1.02 | |
Liquid |
RUO – Research Use Only