missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylammonium hexafluorophosphate, 98%

Product Code. 11317168
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11317168 10 g 10g
11307168 2 g 2g
11327168 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11317168 Supplier Thermo Scientific Alfa Aesar Supplier No. 016256.09
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 429-07-2 | C8H20F6NP | 275.219 g/mol

It is used extensively in electrolytes for electrochemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 429-07-2
Molecular Formula C8H20F6NP
Molecular Weight (g/mol) 275.219
MDL Number MFCD00043179
InChI Key KLKUOIXSIDDDCN-UHFFFAOYSA-N
Synonym tetraethylammonium hexafluorophosphate, ammonium, tetraethyl-, hexafluorophosphate 1-, et4npf6, n,n,n-triethylethanaminium hexafluorophosphate, tetraethylammoniumhexafluorophosphate, tetraethylazanium hexafluorophosphate, tetraethylammonium hexafluorophosphate v, ethanaminium,n,n-triethyl-, hexafluorophosphate 1-, ethanaminium, n,n,n-triethyl-, hexafluorophosphate 1-
PubChem CID 9885
IUPAC Name tetraethylazanium;hexafluorophosphate
SMILES CC[N+](CC)(CC)CC.F[P-](F)(F)(F)(F)F

Specifications

Quantity 10 g
UN Number UN2811
Sensitivity Hygroscopic
Solubility Information Soluble in water,alcohol,and acetonitrile.
Formula Weight 275.23
Percent Purity 98%
Physical Form Crystalline
Chemical Name or Material Tetraethylammonium hexafluorophosphate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.