missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylammonium fluoride hydrate, 97%

Product Code. p-7026884
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11490757 5 g 5g
11400767 25 g 25g
11410767 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11490757 Supplier Thermo Scientific Alfa Aesar Supplier No. B23173.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 98330-04-2 | C8H20FN | 149.25 g/mol

Tetraethylammonium fluoride hydrate is used as a catalyst for oxidation, methylation and deuteration reactions of fluorine. It is involved as a reagent for the cleavage of Si-C bonds and also acts as a phase transfer catalyst in synthetic chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetraethylammonium fluoride hydrate is used as a catalyst for oxidation, methylation and deuteration reactions of fluorine. It is involved as a reagent for the cleavage of Si-C bonds and also acts as a phase transfer catalyst in synthetic chemistry.

Solubility
Soluble in water.

Notes
Hygroscopic. Air sensitive. Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98330-04-2
Molecular Formula C8H20FN
Molecular Weight (g/mol) 149.25
MDL Number MFCD00149990
InChI Key QSUJAUYJBJRLKV-UHFFFAOYSA-M
Synonym tetraethylammonium fluoride hydrate, tetraethylammonium hydrate fluoride, tetraethylammoniumfluoridehydrate, acmc-209nvn, et4nf.h2o, tetraethylazanium fluoride hydrate, tetraethyl ammonium fluoride hydrate, ethanaminium, n,n,n-triethyl-, fluoride, monohydrate
PubChem CID 16211652
IUPAC Name tetraethylazanium;fluoride;hydrate
SMILES [F-].CC[N+](CC)(CC)CC

Specifications

Quantity 5 g
Beilstein 4152659
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 149.26 (Anhydrous)
Percent Purity 97%
Chemical Name or Material Tetraethylammonium fluoride hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.