missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylammonium chloride

Product Code. 15494879
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15494879 25 g 25g
15404889 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15494879 Supplier Thermo Scientific Alfa Aesar Supplier No. J62507.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Phase transfer catalyst | CAS: 56-34-8 | C8H20ClN | 165.71 g/mol

Tetraethylammonium chloride is used as a catalyst. Tetraethylammonium Chloride can be used as a source of tetraethylammonium ions for various pharmaceutical studies and has the ability to block K+ channels in various tissues. Tetraethylammonium Chloride can also block the transmission of nervous impulses across autonomic ganglions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetraethylammonium chloride is used as a catalyst. Tetraethylammonium Chloride can be used as a source of tetraethylammonium ions for various pharmaceutical studies and has the ability to block K+ channels in various tissues. Tetraethylammonium Chloride can also block the transmission of nervous impulses across autonomic ganglions.

Solubility
Soluble in water, ethanol, chloroform, or acetone

Notes
Hygroscopic, Incompatible with strong oxidizing agents. Protect from moisture.  Keep container tightly closed in a dry and well-ventilated place. Handle under argon. Store in cool place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-34-8
Molecular Formula C8H20ClN
Molecular Weight (g/mol) 165.71
MDL Number MFCD00149992
InChI Key YMBCJWGVCUEGHA-UHFFFAOYSA-M
Synonym tetraethylammonium chloride, tetraethyl ammonium chloride, etamon chloride, tea chloride, teac, n,n,n-triethylethanaminium chloride, tetraethylammoniumchloride, ethanaminium, n,n,n-triethyl-, chloride, tetraethyl-ammonium chloride, unii-8b82z83xnn
PubChem CID 5946
ChEBI CHEBI:78161
IUPAC Name tetraethylazanium;chloride
SMILES [Cl-].CC[N+](CC)(CC)CC

Specifications

Color White to Yellow
Linear Formula (CH3CH2)4NCl
Quantity 25 g
Beilstein 3563247
Sensitivity Hygroscopic
Merck Index 14,9200
Solubility Information Soluble in water,ethanol,chloroform,or acetone
Formula Weight 165.71
Physical Form Powder
Chemical Name or Material Tetraethylammonium chloride
compliance-icons
Product Identifier
  • Tetraethylammonium chloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Tetraethylammonium chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.