missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylammonium chloride hydrate, 98%

Product Code. 11462238
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11462238

Brand: Thermo Scientific Alfa Aesar A15215.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 68696-18-4 | C8H22ClNO | 183.72 g/mol

Tetraethylammonium chloride monohydrate is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetraethylammonium chloride monohydrate is used as pharmaceutical intermediate.

Solubility
Soluble in water (1410 g/L at 20°C).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68696-18-4
Molecular Formula C8H22ClNO
Molecular Weight (g/mol) 183.72
MDL Number MFCD00149992
InChI Key CALLTGJPWMIDPC-UHFFFAOYSA-M
Synonym tetraethylammonium chloride hydrate, tetraethylammonium chloride monohydrate, tetraethylazanium chloride hydrate, tetraethylammonium hydrate chloride, c8h20n.cl.h2o, tetraethylammoniumchloride hydrate, tetraethyl ammonium chloride hydrate, tetraethylammonium chloride hydrate, cryst, n,n,n-triethylethanaminium chloride-water 1/1/1, tetraethylammonium chloride, monohydrate
PubChem CID 16211205
IUPAC Name tetraethylazanium;chloride;hydrate
SMILES CC[N+](CC)(CC)CC.O.[Cl-]

Specifications

Melting Point ∼110°C
Density 1.0801
Quantity 500 g
Beilstein 3563247
Merck Index 14,9200
Solubility Information Soluble in water (1410g/L at 20°C).
Formula Weight 183.72 (165.71 Anhydrous)
Percent Purity 98%
Chemical Name or Material Tetraethylammonium chloride monohydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.