missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylammonium chloride hydrate, 98%

Product Code. 11442238
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11442238 25 g 25g
11452238 100 g 100g
11462238 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11442238 Supplier Thermo Scientific Alfa Aesar Supplier No. A15215.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 68696-18-4 | C8H22ClNO | 183.72 g/mol

Tetraethylammonium chloride monohydrate is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetraethylammonium chloride monohydrate is used as pharmaceutical intermediate.

Solubility
Soluble in water (1410 g/L at 20°C).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68696-18-4
Molecular Formula C8H22ClNO
Molecular Weight (g/mol) 183.72
MDL Number MFCD00149992
InChI Key CALLTGJPWMIDPC-UHFFFAOYSA-M
Synonym tetraethylammonium chloride hydrate, tetraethylammonium chloride monohydrate, tetraethylazanium chloride hydrate, tetraethylammonium hydrate chloride, c8h20n.cl.h2o, tetraethylammoniumchloride hydrate, tetraethyl ammonium chloride hydrate, tetraethylammonium chloride hydrate, cryst, n,n,n-triethylethanaminium chloride-water 1/1/1, tetraethylammonium chloride, monohydrate
PubChem CID 16211205
IUPAC Name tetraethylazanium;chloride;hydrate
SMILES CC[N+](CC)(CC)CC.O.[Cl-]

Specifications

Melting Point ∼110°C
Density 1.0801
Quantity 25 g
Beilstein 3563247
Merck Index 14,9200
Solubility Information Soluble in water (1410g/L at 20°C).
Formula Weight 183.72 (165.71 Anhydrous)
Percent Purity 98%
Chemical Name or Material Tetraethylammonium chloride monohydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.