missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylammonium Bromide, TRC

Product Code. 30416214
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30416214 50 g 50g
30414664 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30416214 Supplier Toronto Research Chemicals Supplier No. TRCT29220550G
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tetraethylammonium Bromide
CAS 71-91-0
Molecular Formula C8H20BrN
Quantity 50 g
Synonym N,N,N-triethylehanaminium Bromide (1:1), Beparon, Bromethyl, Etambro, Etamon, Etylon, Sympatektoman, TEA bromide, TEAB, TMD 10, Teamon, Tetranium, Tetrylammonium bromide
InChI Formula InChI=1S/C8H20N.BrH/c1-5-9(6-2,7-3)8-4;/h5-8H2,1-4H3;1H/q+1;/p-1
SMILES [Br-].CC[N+](CC)(CC)CC
IUPAC Name tetraethylazanium;bromide
Molecular Weight (g/mol) 210.16
Formula Weight 209.0779
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.