missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetraethylammonium bromide, 99+%, Thermo Scientific Chemicals
29.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
50 g
250 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic bottle
Descripción
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
Especificaciones
Melting Point | 285.0°C |
Color | White to Cream |
Loss on Drying | 1.0% max. |
Infrared Spectrum | Authentic |
Assay Percent Range | 99% min. (Argentometry) |
Packaging | Glass bottle |
Linear Formula | (C2H5)4NBr |
Quantity | 50 g |
Beilstein | 04,104 |
Fieser | 06,568; 13,46 |
Mostrar más |
Seguridad y manipulación
Clasificación
- LONG-TERM AQUATIC HAZARD Chronic 3
- H412-Harmful to aquatic life with long lasting effects.
- P273-Avoid release to the environment.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'