missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylammonium Bromide, 98+%, Extra Pure, SLR, Fisher Chemical

Product Code. 10000360 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Plastic powder jar
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10000360 100 g Plastic powder jar 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10000360 Supplier Fisher Chemical Supplier No. T/0620/48

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Melting Point 285°C
CAS 71-91-0
Color Cream
pH 6.5
Physical Form Powder/Solid
Quantity 100 g
Molecular Formula C8H20BrN
MDL Number 11825
Synonym tetraethylammonium bromide, teab, tea bromide, tetrylammonium bromide, sympatektoman, tetranium, etylon, bromethyl, beparon, etambro
InChI Key HWCKGOZZJDHMNC-UHFFFAOYSA-M
SMILES CC[N+](CC)(CC)CC.[Br-]
IUPAC Name tetraethylazanium;bromide
Molecular Weight (g/mol) 210.159
PubChem CID 6285
Formula Weight 210.16g/mol
Packaging Plastic powder jar
Show More Show Less
Product Identifier
  • Tetraethylammonium bromide
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.