missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylammonium acetate tetrahydrate, 97+%, Thermo Scientific Chemicals

Catalog Number 10656022
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
67533-12-4
C10H23NO2·4H2O
261.35
QNOAKQGNSWGYOE-UHFFFAOYSA-M
tetraethylammonium acetate tetrahydrate, ethanaminium, n,n,n-triethyl-, acetate, tetrahydrate, tetraethylazanium acetate tetrahydrate, tetraethylazanium ethanoate tetrahydrate, acetate ion tetraethylammonium tetrahydrate, ethanaminium,n,n,n-triethyl-,acetate,tetrahydrate
2724098
tetraethylazanium;acetate;tetrahydrate
CC[N+](CC)(CC)CC.CC(=O)[O-].O.O.O.O
This item is not returnable. View return policy
67533-12-4
C10H23NO2·4H2O
261.35
QNOAKQGNSWGYOE-UHFFFAOYSA-M
tetraethylammonium acetate tetrahydrate, ethanaminium, n,n,n-triethyl-, acetate, tetrahydrate, tetraethylazanium acetate tetrahydrate, tetraethylazanium ethanoate tetrahydrate, acetate ion tetraethylammonium tetrahydrate, ethanaminium,n,n,n-triethyl-,acetate,tetrahydrate
2724098
tetraethylazanium;acetate;tetrahydrate
CC[N+](CC)(CC)CC.CC(=O)[O-].O.O.O.O

Phase transfer catalyst

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY
Melting Point 40.0°C to 45.0°C
Color Colorless to White
Flash Point 60°C
Infrared Spectrum Authentic
Assay Percent Range 97+%
Packaging Glass bottle
Linear Formula (C2H5)4N(O2CCH3)·4H2O
Quantity 25 g
Solubility Information Solubility in water: soluble.
Formula Weight 261.35
Percent Purity 97+%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Tetraethylammonium acetate tetrahydrate
Show More Show Less
compliance-icons
Product Identifier
  • Tetraethylammonium acetate tetrahydrate
Signal Word
  • Danger
Hazard Category
  • Flammable solid Category 1
Hazard Statement
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'