missing translation for 'onlineSavingsMsg'
Learn More

Tetraethyl orthosilicate, AcroSeal™, ACROS Organics™

Brand:  Acros Organics 420361000

Code : 29

Additional Details : CAS Number : 78-10-4 Weight : 0.14000kg Transport : UN number : 1292 Chem class : 1 Pack group : III


Quantity 100mL
Packaging AcroSeal™ Glass Bottle

 View more versions of this product

Product Code. 10195897

  • £16.30 / 100mL


EU Stock 19
Add to basket


Specifications

Chemical Identifiers

Tetraethyl orthosilicate
98%
Si(OC2H5)4
Ethyl silicate
98%
C8H20O4Si
208.33

Specifications

Tetraethyl orthosilicate
45°C
166.0° to 169.0°C
− 77.0°C
78-10-4
Authentic
1.3820 to 1.3840
Liquid
98%
Si(OC2H5)4
Ethyl silicate
0.9400g/mL
0.94
Clear
100mL
1% max.
AcroSeal™ Glass Bottle
Solubility in water: hydrolysis. Other solubilities: miscible with alcohol, hydrocarbons, aromatics,, esters, ketones,, slightly soluble in benzene
98%
C8H20O4Si
208.33
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Tetraethyl orthosilicate

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS