missing translation for 'onlineSavingsMsg'
Learn More

Tetraethyl Fluorescein Bis(methylene)bis(azanetriyl)tetraacetate, TRC

Product Code. 30415790
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30415790 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30415790 Supplier Toronto Research Chemicals Supplier No. TRCT292720100MG
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1170856-93-5
Molecular Formula C38H42N2O13
Quantity 100 mg
Synonym Tetraethyl 2,2’,2’’,2’’’-(((3’,6’-Dihydroxy-3-oxo-3H-spiro[isobenzofuran-1,9’-xanthene]-4’,5’-diyl)bis(methylene))bis(azanetriyl))tetraacetate
InChI Formula InChI=1S/C38H42N2O13/c1-5-48-31(43)19-39(20-32(44)49-6-2)17-24-29(41)15-13-27-35(24)52-36-25(18-40(21-33(45)50-7-3)22-34(46)51-8-4)30(42)16-14-28(36)38(27)26-12-10-9-11-23(26)37(47)53-38/h9-16,41-42H,5-8,17-22H2,1-4H3
SMILES CCOC(=O)CN(CC(=O)OCC)Cc1c(O)ccc2c1Oc3c(CN(CC(=O)OCC)CC(=O)OCC)c(O)ccc3C24OC(=O)c5ccccc45
IUPAC Name ethyl 2-[[5'-[[bis(2-ethoxy-2-oxoethyl)amino]methyl]-3',6'-dihydroxy-3-oxospiro[2-benzofuran-1,9'-xanthene]-4'-yl]methyl-(2-ethoxy-2-oxoethyl)amino]acetate
Molecular Weight (g/mol) 734.75
Formula Weight 734.2687
Chemical Name or Material Tetraethyl Fluorescein Bis(methylene)bis(azanetriyl)tetraacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.