missing translation for 'onlineSavingsMsg'
Learn More

Tetraethyl Ethylenephosphonate, TRC

Product Code. 30421100
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30421100 500 mg 500mg
30415020 100 mg -
30409125 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30421100 Supplier Toronto Research Chemicals Supplier No. TRCT204628500MG
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 995-32-4
Molecular Formula C10H24O6P2
Molecular Weight (g/mol) 302.24
Synonym Phosphonic acid, P,P'-1,2-ethanediylbis-, P,P,P',P'-tetraethyl ester, Phosphonic acid, 1,2-ethanediylbis-, tetraethyl ester (9CI), Phosphonic acid, ethylenedi-, tetraethyl ester (6CI,7CI,8CI), 1,2-Bis(diethoxyphosphinyl)ethane, 1,2-Bis(diethoxyphosphoryl)ethane, 1,2-Bis(diethylphosphonyl)ethane, 1,2-Ethylenediphosphonic acid tetraethyl ester, Ethane-1,2-diphosphonic acid tetraethyl ester, Tetraethyl ethylenebis(phosphonate), Tetraethyl ethylenediphosphonate
IUPAC Name 1,2-bis(diethoxyphosphoryl)ethane
SMILES CCOP(=O)(CCP(=O)(OCC)OCC)OCC

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C10H24O6P2/c1-5-13-17(11,14-6-2)9-10-18(12,15-7-3)16-8-4/h5-10H2,1-4H3
Formula Weight 302.1048
Chemical Name or Material Tetraethyl Ethylenephosphonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.