missing translation for 'onlineSavingsMsg'
Learn More

Tetraethoxysilane, 99+%

Product Code. p-7082605
Change view
Click to view available options
Quantity:
2.5 kg
4 x 500 g
Unit Size:
2.5kg
2000g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11377687 4 x 500 g 2000g
11387687 2.5 kg 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11377687 Supplier Thermo Scientific Alfa Aesar Supplier No. 014082.57
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

As a binding agent and as a polymerization catalyst | CAS: 78-10-4 | C8H20O4Si | 208.329 g/mol

As a binding agent and as a polymerization catalystTetraethoxysilane is the main precursor material for the synthesis of zeolites and silicon dioxide, which is used in semiconductor industry. It is widely used as a cross linking agent in silicon polymers and in aerogel preparations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-10-4
Molecular Formula C8H20O4Si
Molecular Weight (g/mol) 208.329
MDL Number MFCD00009062
InChI Key BOTDANWDWHJENH-UHFFFAOYSA-N
Synonym tetraethyl orthosilicate, tetraethoxysilane, teos, ethyl silicate, silicon ethoxide, silicon tetraethoxide, silane, tetraethoxy, dynasil a, tetraethoxysilicon, ethyl orthosilicate
PubChem CID 6517
IUPAC Name tetraethyl silicate
SMILES CCO[Si](OCC)(OCC)OCC

Specifications

Boiling Point 168°C
Melting Point -85°C
Color Colorless
Physical Form Liquid
Quantity 4 x 500 g
Linear Formula (C2H5O)4Si
UN Number UN1292
Beilstein 1422225
Flash Point 46°C (116°F)
Merck Index 14,3851
Solubility Information Soluble in ethanol and 2-propanol.
Formula Weight 208.33
Percent Purity ≥99%
Odor Pleasant
Refractive Index 1.382
Sensitivity Moisture sensitive
Density 0.934
Chemical Name or Material Tetraethoxysilane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.