missing translation for 'onlineSavingsMsg'
Learn More

Tetradecyltrimethylammonium Bromide, for HPLC, Certified, Fisher Chemical

Product Code. 10743811 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Plastic powder jar
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10743811 25 g Plastic powder jar 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10743811 Supplier Fisher Chemical Supplier No. T/0611/46

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Melting Point 245°C
CAS 1119-97-7
Color White
pH 6 to 8
Physical Form Powder/Solid
Quantity 25 g
Molecular Formula C17H38BrN
MDL Number MFCD00011770
Synonym cetrimide, tetradecyltrimethylammonium bromide, myristyltrimethylammonium bromide, tetradonium bromide, n,n,n-trimethyltetradecan-1-aminium bromide, tetradecyl trimethyl ammonium bromide, myrtrimonium bromide, mytab, mitmab, morpan t
InChI Key CXRFDZFCGOPDTD-UHFFFAOYSA-M
SMILES [Br-].CCCCCCCCCCCCCC[N+](C)(C)C
IUPAC Name trimethyl(tetradecyl)azanium bromide
Molecular Weight (g/mol) 336.40
PubChem CID 14250
ChEBI CHEBI:3565
Formula Weight 336.39g/mol
Packaging Plastic powder jar
Show More Show Less
compliance-icons
Product Identifier
  • Myristyltrimethylammonium bromide
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.