missing translation for 'onlineSavingsMsg'
Learn More

Tetradecyltrimethyl-Ammonium Bromide, 98%, MP Biomedicals™

Product Code. 11494451 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Unit Size:
100g
1kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11494451 1 kg 1kg
17811381 100 g 100g
11484451 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11494451 Supplier MP Biomedicals Supplier No. 0215210991

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 1119-97-7
Molecular Formula C17H38BrN
Molecular Weight (g/mol) 336.40
MDL Number MFCD00011770
InChI Key CXRFDZFCGOPDTD-UHFFFAOYSA-M
Synonym Myristyltrimethylammonium bromide, Trimethyl(tetradecyl)ammonium bromide
PubChem CID 14250
ChEBI CHEBI:3565
SMILES [Br-].CCCCCCCCCCCCCC[N+](C)(C)C

Specifications

Physical Form Powder
Quantity 1 kg
Assay Percent Range 98%
IUPAC Name trimethyl(tetradecyl)azanium bromide
Formula Weight 336.4
Chemical Name or Material Tetradecyltrimethyl-Ammonium Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.