missing translation for 'onlineSavingsMsg'
Learn More

Tetradecanoic acid, 98%

Product Code. 11469962
Change view
Click to view available options
Quantity:
250 g
500 g
2500 g
Unit Size:
2500g
250g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11469962 2500 g 2500g
11443950 250 g 250g
11459962 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11469962 Supplier Thermo Scientific Alfa Aesar Supplier No. A12067.0E
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 544-63-8 | C14H28O2 | 228.38 g/mol

It is employed as a flow Agent and emulsifier. Used in artificial orris fragrances. Employed as anionic/nonionic Surfactants in soaps and detergents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a flow Agent and emulsifier. Used in artificial orris fragrances. Employed as anionic/nonionic Surfactants in soaps and detergents.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed. Store in cool, dry conditions in well ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 544-63-8
Molecular Formula C14H28O2
Molecular Weight (g/mol) 228.38
MDL Number MFCD00002744
InChI Key TUNFSRHWOTWDNC-UHFFFAOYSA-N
Synonym myristic acid, n-tetradecanoic acid, n-tetradecoic acid, crodacid, n-tetradecan-1-oic acid, 1-tridecanecarboxylic acid, hydrofol acid 1495, univol u 316s, emery 655, myristinsaeure
PubChem CID 11005
ChEBI CHEBI:28875
IUPAC Name tetradecanoic acid
SMILES CCCCCCCCCCCCCC(O)=O

Specifications

Melting Point 53°C to 56°C
Density 0.862
Boiling Point 326°C
Flash Point 175°C (347°F)
Quantity 2500 g
Beilstein 508624
Merck Index 14,6333
Solubility Information Insoluble in water.
Formula Weight 228.38
Percent Purity 98%
Chemical Name or Material Tetradecanoic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.