missing translation for 'onlineSavingsMsg'
Learn More

Tetradecanedioic acid, 98%

Product Code. 15496728
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15496728 25 g 25g
15486728 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15496728 Supplier Thermo Scientific Alfa Aesar Supplier No. H60254.14
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 821-38-5 | C14H26O4 | 258.358 g/mol

Tetradecanedioic acid is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetradecanedioic acid is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents, reducing agents amnd bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 821-38-5
Molecular Formula C14H26O4
Molecular Weight (g/mol) 258.358
MDL Number MFCD00002743
InChI Key HQHCYKULIHKCEB-UHFFFAOYSA-N
Synonym 1,12-dodecanedicarboxylic acid, tetradecane-1,14-dioic acid, 1,14-tetradecanedioic acid, unii-uec3ac47h1, dodecamethylenedicarboxylic acid, tetradecanedicarboxylic acid, uec3ac47h1, 1,12-dodecanedicarboxylicacid, tetradecanedicarboxylate, pubchem18099
PubChem CID 13185
ChEBI CHEBI:76308
IUPAC Name tetradecanedioic acid
SMILES C(CCCCCCC(=O)O)CCCCCC(=O)O

Specifications

Melting Point 124°C to 127°C
Linear Formula HOOC(CH2)12COOH
Quantity 25 g
Beilstein 1788202
Solubility Information Slightly soluble in water.
Formula Weight 258.35
Percent Purity 98%
Chemical Name or Material Tetradecanedioic acid
compliance-icons
Product Identifier
  • Tetradecanedioic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Tetradecanedioic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.