missing translation for 'onlineSavingsMsg'
Learn More

Tetracyclohexyltin

Product Code. p-7049701
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11363487

Brand: Thermo Scientific Alfa Aesar 018822.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1449-55-4 | C24H44Sn | 451.326 g/mol

It is used as the ion carrier for the construction of a highly selective salicylate sensor. This sensor shows a Nernstian response to salicylate ions over a very wide concentration (1.0 × 10-7-1.0 × 10-1 M) in a pH range of 5.5-10.5.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1449-55-4
Molecular Formula C24H44Sn
Molecular Weight (g/mol) 451.326
MDL Number MFCD00014278
InChI Key JUISPCSEIXBMNI-UHFFFAOYSA-N
Synonym tetracyclohexyltin, stannane, tetracyclohexyl, tetracyclohexylstananne, tetracyclohexyl tin, tin, tetracyclohexyl, acmc-1c0rn
PubChem CID 74056
IUPAC Name tetracyclohexylstannane
SMILES C1CCC(CC1)[Sn](C2CCCCC2)(C3CCCCC3)C4CCCCC4

Specifications

Melting Point 241°C to 250°C
Quantity 1 g
UN Number UN3146
Solubility Information Sparingly Soluble (8.1E-9 g/L) (25°C),
Formula Weight 451.31
Physical Form Powder
Chemical Name or Material Tetracyclohexyltin

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.