missing translation for 'onlineSavingsMsg'
Learn More

Tetracyclohexyltin

Product Code. 11383487
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11383487 25 g 25g
11363487 1 g 1g
11373487 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11383487 Supplier Thermo Scientific Alfa Aesar Supplier No. 018822.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1449-55-4 | C24H44Sn | 451.326 g/mol

It is used as the ion carrier for the construction of a highly selective salicylate sensor. This sensor shows a Nernstian response to salicylate ions over a very wide concentration (1.0 × 10-7-1.0 × 10-1 M) in a pH range of 5.5-10.5.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1449-55-4
Molecular Formula C24H44Sn
Molecular Weight (g/mol) 451.326
MDL Number MFCD00014278
InChI Key JUISPCSEIXBMNI-UHFFFAOYSA-N
Synonym tetracyclohexyltin, stannane, tetracyclohexyl, tetracyclohexylstananne, tetracyclohexyl tin, tin, tetracyclohexyl, acmc-1c0rn
PubChem CID 74056
IUPAC Name tetracyclohexylstannane
SMILES C1CCC(CC1)[Sn](C2CCCCC2)(C3CCCCC3)C4CCCCC4

Specifications

Melting Point 241°C to 250°C
Quantity 25 g
UN Number UN3146
Solubility Information Sparingly Soluble (8.1E-9 g/L) (25°C),
Formula Weight 451.31
Physical Form Powder
Chemical Name or Material Tetracyclohexyltin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.