missing translation for 'onlineSavingsMsg'
Learn More

Tetracycline Hydrochloride, TRC

Product Code. 30009158
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30009158 5 g 5g
30005861 10 g 10g
17984285 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009158 Supplier Toronto Research Chemicals Supplier No. TRCT2914005G
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-75-5
Molecular Formula C22 H24 N2 O8 . Cl H
Molecular Weight (g/mol) 480.9
Synonym Tetracycline hydrochloride, Oxytetracycline Hydrochloride Imp. B (EP) as Hydrochloride, (4S,4aS,5aS,6S,12aS)-4-(Dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,6,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-2-naphthacenecarboxamide monohydrochloride, 4-(Dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,6,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-2-naphthacenecarboxamide monohydrochloride, [4S-(4α,4aα,5aα,6β,12aα)]-4-(Dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,6,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-2-Naphthacenecarboxamide monohydrochloride, 2-Naphthacenecarboxamide, 4-(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,6,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-, hydrochloride (1:1), (4S,4aS,5aS,6S,12aS)-, 2-Naphthacenecarboxamide, 4-(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,6,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-, monohydrochloride (8CI), 2-Naphthacenecarboxamide, 4-(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,6,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-, monohydrochloride, (4S,4aS,5aS,6S,12aS)- (9CI), 2-Naphthacenecarboxamide, 4-(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,6,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-, monohydrochloride, [4S-(4α,4aα,5aα,6β,12aα)]-, (-)-Tetracycline hydrochloride, Achro, Achromycin hydrochloride, Ala-Tet, Ambracyn, Bristaciclina, Bristacycline, Cancycline-250, Cefracycline, Criseociclina, Cyclopar, Diocyclin, Dumocyclin, Helvecyclin, Heromycin, Hostacyclin, Hostacycline, Imex, Laudin, Medamycin, Neocycline B, Paltet, Panmycin hydrochloride, Partrex, Polycycline hydrochloride, Polyotic ointment, Purocyclina, Quadracyclin, Remicyclin, Riocyclin, Ro-Cycline, Robitet, Sanclomycine, Steclin, Subamycin, Supramycin, Sustamycin, Talcin, Tefilin, Tet-Cy, TetraSURE, Tetrabakat, Tetrabid, Tetrablet, Tetrabon, Tetracaps, Tetrachel, Tetracompren, Tetracycline chlorohydrate, Tetracyklin, Tetracyn, Tetrakap, Tetralution, Tetramavan, Tetrosol, Topicycline, Triphacyclin, Unicin, Vetquamycin 324
IUPAC Name (4S,4aS,5aS,6S,12aS)-4-(dimethylamino)-3,6,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride
SMILES Cl.CN(C)[C@H]1[C@@H]2C[C@H]3C(=C(O)[C@]2(O)C(=O)C(=C1O)C(=O)N)C(=O)c4c(O)cccc4[C@@]3(C)O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C22H24N2O8.ClH/c1-21(31)8-5-4-6-11(25)12(8)16(26)13-9(21)7-10-15(24(2)3)17(27)14(20(23)30)19(29)22(10,32)18(13)28;/h4-6,9-10,15,25,27-28,31-32H,7H2,1-3H3,(H2,23,30);1H/t9-,10-,15-,21+,22-;/m0./s1
Formula Weight 480.1299
Percent Purity >95%
Chemical Name or Material Tetracycline Hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.