missing translation for 'onlineSavingsMsg'
Learn More

Tetracycline hydrochloride, MedChemExpress

Product Code. 16482189
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16482189 1 g 1g
16460948 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16482189 Supplier Medchem Express Supplier No. HYB0474
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Tetracycline (hydrochloride) is a broad-spectrum antibiotic, exhibiting activity against a wide range of gram-positive and gram-negative bacteria.

Tetracycline (hydrochloride) is a broad-spectrum antibiotic, exhibiting activity against a wide range of gram-positive and gram-negative bacteria.

Specifications

Chemical Name or Material Tetracycline hydrochloride
Physical Form Powder
CAS 64-75-5
Color Yellow
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C22H25ClN2O8
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 62.5 mg/mL (129.96 mM; Need ultrasonic) ∣H2O : 50 mg/mL (103.97 mM; Need ultrasonic)
SMILES O[C@@]1(C(C(C(N)=O)=C2O)=O)[C@@]([C@@H]2N(C)C)([H])C[C@@]([C@](O)(C3=CC=C4)C)([H])C(C(C3=C4O)=O)=C1O.Cl
Molecular Weight (g/mol) 480.9
Formula Weight 480.9
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.