missing translation for 'onlineSavingsMsg'
Learn More

Tetracycline Hydrochloride, LoGiCal

Product Code. 30461503
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30461503 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30461503 Supplier LoGiCal Supplier No. LGCFOR0452.00
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tetracycline Hydrochloride
CAS 64-75-5
Analyte or Component Names Tetracycline Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C22 H24 N2 O8 . Cl H
Certifications/Compliance ISO 9001
Synonym Oxytetracycline Hydrochloride Imp. B (EP), Oxytetracycline Imp. B (EP), Tetracycline Hydrochloride, Oxytetracycline Hydrochloride Imp. B (EP) as Hydrochloride, (4S,4aS,5aS,6S,12aS)-4-(Dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,6,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-2-naphthacenecarboxamide monohydrochloride, 4-(Dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,6,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-2-naphthacenecarboxamide monohydrochloride, [4S-(4alpha,4aalpha,5aalpha,6beta,12aalpha)]-4-(Dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,6,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-2-Naphthacenecarboxamide monohydrochloride, (-)-Tetracycline hydrochloride, Achro, Achromycin hydrochloride, Ala-Tet, Ambracyn, Bristaciclina, Bristacycline, Cancycline-250, Cefracycline, Criseociclina, Cyclopar, Diocyclin, Dumocyclin, Helvecyclin, Hostacyclin, Hostacycline, Imex, Medamycin, Neocycline B, Paltet, Panmycin hydrochloride, Partrex, Polycycline hydrochloride, Polyotic ointment, Purocyclina, Quadracyclin, Remicyclin, Riocyclin, Ro-Cycline, Robitet, Sanclomycine, Steclin, Subamycin, Supramycin, Sustamycin, Tefilin, Tet-Cy, TetraSURE, Tetrabakat, Tetrabid, Tetrablet, Tetrabon, Tetracaps, Tetrachel, Tetracompren, Tetracycline chlorohydrate, Tetracycline hydrochloride, Tetracyklin, Tetracyn, Tetrakap, Tetralution, Tetramavan, Tetrosol, Topicycline, Totomycin, Triphacyclin, Unicin, Vetquamycin 324, Oxytetracycline Hydrochloride Impurity B as Hydrochloride, Oxytetracycline Impurity B as Hydrochloride
InChI Formula InChI=1S/C22H24N2O8.ClH/c1-21(31)8-5-4-6-11(25)12(8)16(26)13-9(21)7-10-15(24(2)3)17(27)14(20(23)30)19(29)22(10,32)18(13)28;/h4-6,9-10,15,25,27-28,31-32H,7H2,1-3H3,(H2,23,30);1H/t9-,10-,15-,21+,22-;/m0./s1
Solution Type Neat
SMILES Cl.CN(C)[C@H]1[C@@H]2C[C@H]3C(=C(O)[C@]2(O)C(=O)C(=C1O)C(=O)N)C(=O)c4c(O)cccc4[C@@]3(C)O
IUPAC Name (4S,4aS,5aS,6S,12aS)-4-(dimethylamino)-3,6,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride
Industry Type Clinical Testing
Molecular Weight (g/mol) 480.9
Quantity 10 mg
Formula Weight 480.1299 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.