missing translation for 'onlineSavingsMsg'
Learn More

Tetracycline hydrochloride, 96%

Codice prodotto. 11479367
Cambia vista
Click to view available options
Quantity:
25 g
100 g
Dimensione della confezione:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantity Dimensione della confezione
11479367 25 g 25g
11489367 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 11479367 Fornitore Thermo Scientific Alfa Aesar N. del fornitore B21408.14
 Restricted Item
In Stock
Aggiungi al carrello
Aggiungi al carrello
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Tetracycline hydrochloride, CAS # 64-75-5, is a wide-spectrum antibiotic that targets both gram-negative and gram-positive bacteria. | CAS: 64-75-5 | C22H25ClN2O8 | 480.898 g/mol

Tetracycline hydrochloride is used to induce apoptosis in osteoclasts. It is used to treat acne and other skin infections, respiratory tract infections like pneumonia, genital, urinary infections, leptospirosis, helicobacter pylori, taxoplasmosis, mycoplasma, psittacosis for dog and cats. It also works more effectively in animals that have tick-borne infections. It is also useful in cell culture applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Tetracycline hydrochloride is the hydrochloride salt of the antibiotic tetracycline
  • Tetracycline blocks amino acyl-tRNA binding during bacterial protein synthesis
  • Tetracycline is a broad-spectrum naphthacene antibiotic that inhibits growth of both gram positive and gram negative bacteria

Applications

  • Tetracycline may be used as a selection agent in cell culture
  • It may also be used to activate inducible gene expression in engineered cells
TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 64-75-5
Molecular Formula C22H25ClN2O8
Molecular Weight (g/mol) 480.898
MDL Number MFCD00078142
InChI Key HTXDZWDXSWLLLW-FMZCEJRJSA-N
PubChem CID 129628373
IUPAC Name (4S,4aS,5aS,6S,12aS)-4-(dimethylamino)-3,6,10,11,12a-pentahydroxy-6-methyl-1,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride
SMILES CC1(C2CC3C(C(=C(C(=O)C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)C(=O)N)O)N(C)C)O.Cl

Especificaciones

Type Tetracycline Hydrochloride
Melting Point 220°C to 223°C
Quantity 25 g
Assay Percent Range 0.98
Sensitivity Air and light sensitive
Merck Index 14,9196
Solubility Information Soluble in water,dimethyl sulfoxide,methanol and ethanol. Insoluble in ether and hydrocarbons.
Formula Weight 480.91
Percent Purity 96%
Chemical Name or Material Tetracycline hydrochloride
compliance-icons compliance-icons
Nombre del producto
  • Tetracycline hydrochloride
Palabra de advertencia
  • Warning
Clasificación
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 2
Indicaciones de peligro
  • H319-Causes serious eye irritation.
  • H361d-Suspected of damaging the unborn child.
Consejos de prudencia
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Otros peligros
  • MIXTURE LIST-Contain: Tetracycline hydrochloride

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.