missing translation for 'onlineSavingsMsg'
Learn More

Tetracyanoethylene, 98%

Product Code. p-4509931 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
10g
25g
This item is not returnable. View return policy

Product Code. 10316450

Brand: Thermo Scientific Acros 138050100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 670-54-2 | C6N4 | 128.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent, Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 670-54-2
Molecular Formula C6N4
Molecular Weight (g/mol) 128.09
InChI Key NLDYACGHTUPAQU-UHFFFAOYSA-N
Synonym tetracyanoethylene, tetracyanoethene, ethylenetetracarbonitrile, tcne, ethenetetracarbonitrile, ethene, tetracyano, 1,1,2,2-ethenetetracarbonitrile, tetrakyanethylen, 1,1,2,2-tetracyanoethene, 1,1,2,2-tetracyanoethylene
PubChem CID 12635
IUPAC Name ethene-1,1,2,2-tetracarbonitrile
SMILES C(#N)C(=C(C#N)C#N)C#N

Specifications

Melting Point 196.0°C to 201.0°C
Boiling Point 223.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Ex N)
Packaging Glass bottle
Linear Formula (NC)2C=C(CN)2
Quantity 10 g
Fieser 01,1133; 02,397; 05,647; 06,567; 13,289
Merck Index 15,934
Solubility Information Solubility in water: hydrolyzes (forming hcn) in water. Other solubilities: slightly soluble in: ether, benzene,, carbon tetrachloride, soluble in: acetone
Formula Weight 128.09
Percent Purity 98%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material Tetracyanoethylene
Show More Show Less
compliance-icons
Product Identifier
  • Tetracyanoethylene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H310-Fatal in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P330-Rinse mouth.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.