missing translation for 'onlineSavingsMsg'
Learn More

Tetracyanoethylene, 98%, Thermo Scientific™

Product Code. 17322481 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17322481 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17322481 Supplier Thermo Scientific Acros Supplier No. 138050050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 670-54-2
Melting Point 196°C to 201°C
Boiling Point 223°C (760 mmHg)
Infrared Spectrum Conforms to structure
Assay Percent Range ≥ 97.5% (Ex N)
Packaging Glass Bottle
Molecular Formula C6N4
Linear Formula (NC)2C=C(CN)2
MDL Number 00001850
Quantity 5 g
UN Number 3439
Fieser 13-289
Sensitivity Moisture sensitive
Merck Index 9340
Synonym TCNE
InChI Key NLDYACGHTUPAQU-UHFFFAOYSA-N
SMILES N#CC(C#N)=C(C#N)C#N
IUPAC Name eth-1-ene-1,1,2,2-tetracarbonitrile
Molecular Weight (g/mol) 128.09
Formula Weight 128.09
Percent Purity 98%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material Tetracyanoethylene
Show More Show Less
compliance-icons
Product Identifier
  • Tetracyanoethylene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P330-Rinse mouth.
Supplemental information
  • MIXTURE LIST-Contain: Tetracyanomethylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.