missing translation for 'onlineSavingsMsg'
Learn More

Tetracosanol, 99+%

Product Code. 10114820 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mg
1 g
Packaging:
Glass bottle
Unit Size:
100mg
1g
This item is not returnable. View return policy

Product Code. 10114820

Brand: Thermo Scientific Acros 258150010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 506-51-4 | C24H50O | 354.66 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 506-51-4
Molecular Formula C24H50O
Molecular Weight (g/mol) 354.66
MDL Number MFCD00042662
InChI Key TYWMIZZBOVGFOV-UHFFFAOYSA-N
Synonym 1-tetracosanol, tetracosanol, lignoceryl alcohol, lignocerol, n-tetracosanol, tetracosyl alcohol, lignoceric alcohol, unii-2n0pi37ioc, n-tetracosanol-1, 2n0pi37ioc
PubChem CID 10472
ChEBI CHEBI:77413
IUPAC Name tetracosan-1-ol
SMILES CCCCCCCCCCCCCCCCCCCCCCCCO

Specifications

Melting Point 75°C to 77°C
Color White
Flash Point 218°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula CH3(CH2)23OH
Quantity 1 g
Solubility Information Solubility in water: .. Other solubilities: soluble in hydrocarbons,alcohols,ether,ketones,,specially when hot
Formula Weight 354.65
Percent Purity 99+%
Physical Form Shiny Flakes or Fluffy Powder
Chemical Name or Material Tetracosanol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.