missing translation for 'onlineSavingsMsg'
Learn More

Tetracosanoic Acid, TRC

Product Code. 30009466
Change view
Click to view available options
Quantity:
100 mg
1 g
2.5 g
Unit Size:
100mg
1g
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009466 2.5 g 2.5g
30007583 1 g 1g
30007292 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009466 Supplier Toronto Research Chemicals Supplier No. TRCT2914552.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 557-59-5
Molecular Formula C24 H48 O2
Molecular Weight (g/mol) 368.64
Synonym Tetracosanoic acid (8CI, 9CI, ACI), FL 88, FL 88 (fatty acid), L 88, L 88 (fatty acid), Lignoceric acid, n-Tetracosanoic acid
IUPAC Name tetracosanoic acid
SMILES CCCCCCCCCCCCCCCCCCCCCCCC(=O)O

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C24H48O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-24(25)26/h2-23H2,1H3,(H,25,26)
Formula Weight 368.3654
Chemical Name or Material Lignoceric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.