missing translation for 'onlineSavingsMsg'
Learn More

Tetracosanoic acid, 99%

Product Code. 10624912 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10624912 1 g Glass bottle 1g
10549263 250 mg Glass bottle 250mg
10046253 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10624912 Supplier Thermo Scientific Acros Supplier No. 215640010
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 557-59-5 | C24H48O2 | 368.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 557-59-5
Molecular Formula C24H48O2
Molecular Weight (g/mol) 368.63
MDL Number MFCD00002810
InChI Key QZZGJDVWLFXDLK-UHFFFAOYSA-N
Synonym lignoceric acid, unii-rk3vcw5y1l, n-tetracosanoic acid, rk3vcw5y1l, n-tetracosanoate, lignozerinsaeure, tetracosansaeure, carnaubic acid, tetracosoic acid, tetracosanic acid
PubChem CID 11197
ChEBI CHEBI:28866
IUPAC Name tetracosanoic acid
SMILES CCCCCCCCCCCCCCCCCCCCCCCC(=O)O

Specifications

Melting Point 84.0°C to 86.0°C
Color White
Boiling Point 272.0°C (10.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)22CO2H
Quantity 1 g
Beilstein 02, I, 181
Merck Index 15, 5541
Solubility Information Solubility in water: .. Other solubilities: practically insoluble in ethanol, soluble in hot hydrocarbons and hot ketones
Formula Weight 368.63
Percent Purity 99%
Physical Form Shiny Crystalline Powder or Flakes
Chemical Name or Material Tetracosanoic acid
Show More Show Less
compliance-icons
Product Identifier
  • Tetracosanoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.