missing translation for 'onlineSavingsMsg'
Learn More

Tetracontane, 98%, Thermo Scientific Chemicals

Catalog Number 10155370
Click to view available options
Quantity:
500 mg
2.5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
4181-95-7
C40H82
563.10
MFCD00015267
KUPLEGDPSCCPJI-UHFFFAOYSA-N
n-tetracontane, unii-i79s9ixb9y, alkane c40, i79s9ixb9y, tetracontane, analytical standard, acmc-209jlw, n-tetracontane 500mg, tetracontane gc
20149
tetracontane
CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
4181-95-7
C40H82
563.10
MFCD00015267
KUPLEGDPSCCPJI-UHFFFAOYSA-N
n-tetracontane, unii-i79s9ixb9y, alkane c40, i79s9ixb9y, tetracontane, analytical standard, acmc-209jlw, n-tetracontane 500mg, tetracontane gc
20149
tetracontane
CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 80.0°C to 83.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)38CH3
Quantity 500 mg
Beilstein 01,III,592
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in petroleum ether
Formula Weight 563.1
Percent Purity 98%
Physical Form Powder or Flaky Solid
Chemical Name or Material Tetracontane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'