missing translation for 'onlineSavingsMsg'
Learn More

Tetrachlorophthalic Anhydride, TRC

Product Code. 30425500
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30425500 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30425500 Supplier Toronto Research Chemicals Supplier No. TRCT291510500G
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tetrachlorophthalic Anhydride
CAS 117-08-8
Molecular Formula C8Cl4O3
Quantity 500 g
Synonym Tetrachlorophthalic Anhydride, 1,3-Dioxy-4,5,6,7-tetrachloroisobenzofuran, 3,4,5,6-Tetrachlorophthalic Anhydride, 4,5,6,7-Tetrachloroisobenzofuran-1,3-dione, NSC 1484, Niagathal, Tetrachlorophthalic Acid Anhydride
InChI Formula InChI=1S/C8Cl4O3/c9-3-1-2(8(14)15-7(1)13)4(10)6(12)5(3)11
SMILES Clc1c(Cl)c(Cl)c2C(=O)OC(=O)c2c1Cl
IUPAC Name 4,5,6,7-tetrachloro-2-benzofuran-1,3-dione
Molecular Weight (g/mol) 285.9
Formula Weight 283.8602
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.