missing translation for 'onlineSavingsMsg'
Learn More

Tetrachlorohydroquinone, MP Biomedicals

Product Code. 11449064 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11449064 100 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11449064 Supplier MP Biomedicals Supplier No. 0521865101
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 87-87-6
Molecular Formula C6H2Cl4O2
Quantity 100 g
Synonym tetrachlorohydroquinone, 2,3,5,6-tetrachlorohydroquinone, dihydro-p-chloranil, tetrachloro-p-benzohydroquinone, tetrachlorobenzoquinol, nordrosophilin a, 2,3,5,6-tetrachloro-1,4-benzenediol, perchlorohydroquinone, hydroquinone, tetrachloro, 1,4-benzenediol, 2,3,5,6-tetrachloro
InChI Key STOSPPMGXZPHKP-UHFFFAOYSA-N
SMILES C1(=C(C(=C(C(=C1Cl)Cl)O)Cl)Cl)O
IUPAC Name 2,3,5,6-tetrachlorobenzene-1,4-diol
Molecular Weight (g/mol) 247.88
PubChem CID 66603
ChEBI CHEBI:17042
Formula Weight 247.89
Chemical Name or Material Tetrachlorohydroquinone
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.