missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| Packaging | Narrow Mouth Amber Glass Bottle |
| Quantity | 4 L |
| Specific Gravity | 1L = 1.62kg |
| Formula Weight | 165.83 |
| Grade | BAKER ANALYZED™ Reagent |
| Chemical Name or Material | Tetrachloroethylene |
Safety and Handling
Signal Word
- Warning
- LONG-TERM AQUATIC HAZARD Chronic 2
- Carcinogenicity Category 2
- H351-Suspected of causing cancer.
- H411-Toxic to aquatic life with long lasting effects.
- P273-Avoid release to the environment.
- P281-Use personal protective equipment as required.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P391-Collect spillage.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?