missing translation for 'onlineSavingsMsg'
Learn More

Tetrachloroethylene, 99%, extra pure

Product Code. 10469803 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
500 mL
1 L
2.5 L
10 L
Packaging:
Glass Bottle
Plastic drum
Unit Size:
1 L
10 L
2.5 L
500mL
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

Product Code. 10469803

Brand: Thermo Scientific Acros 138010100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

Tetrachloroethylene, 99%, C2Cl4, CAS Number-127-18-4, ankilostin, carbon dichloride, ethene, tetrachloro, ethylene tetrachloride, perc, perchlorethylene, perchloroethylene, tetrachlorethylene, tetrachloroethene, tetrachloroethylene, 1L, 0.003% max. (NaOH), 98.5% min. (GC), 01, 187 | CAS: 127-18-4 | C2Cl4 | 165.82 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-18-4
Molecular Formula C2Cl4
Molecular Weight (g/mol) 165.82
MDL Number MFCD00000834
InChI Key CYTYCFOTNPOANT-UHFFFAOYSA-N
Synonym tetrachloroethylene, tetrachloroethene, perchloroethylene, perchlorethylene, perc, ethene, tetrachloro, tetrachlorethylene, ethylene tetrachloride, carbon dichloride, ankilostin
PubChem CID 31373
ChEBI CHEBI:17300
IUPAC Name 1,1,2,2-tetrachloroethene
SMILES ClC(Cl)=C(Cl)Cl

Specifications

Alkalinity 0.003% max. (NaOH)
Melting Point -22.0°C
Density 1.6250g/mL
Boiling Point 120.0°C to 122.0°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic drum
Linear Formula Cl2C=CCl2
Refractive Index 1.505
Quantity 10 L
Beilstein 01, 187
Residue after Evaporation 0.005% max.
Merck Index 15, 9335
Specific Gravity 1.625
Solubility Information Solubility in water: 0.15g/L (20°C). Other solubilities: miscible with alcohol, ether, oils, chloroform, and benzene
Viscosity 0.89 mPa.s (20°C)
Formula Weight 165.83
Percent Purity 99%
Grade Extra Pure
Physical Form Liquid
Chemical Name or Material Tetrachloroethylene
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Tetrachloroethylene
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H411-Toxic to aquatic life with long lasting effects.
  • H336-May cause drowsiness or dizziness.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.