missing translation for 'onlineSavingsMsg'
Learn More

Tetrachloroethylene, ≥99.8%, Honeywell™ Riedel-de-Haën™

Product Code. 15637080
Change view
Click to view available options
Quantity:
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15637080 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15637080 Supplier Honeywell Riedel-de Haën Supplier No. 78520100ML
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 127-18-4
Density 1.623g/mL at 25°C
Packaging Glass bottle
Molecular Formula C2Cl4
Linear Formula CCl2=CCl2
MDL Number MFCD00000834
Quantity 100 mL
UN Number UN1897
Beilstein 1361721
Synonym tetrachloroethylene, tetrachloroethene, perchloroethylene, perchlorethylene, perc, ethene, tetrachloro, tetrachlorethylene, ethylene tetrachloride, carbon dichloride, ankilostin
InChI Key CYTYCFOTNPOANT-UHFFFAOYSA-N
SMILES ClC(Cl)=C(Cl)Cl
IUPAC Name 1,1,2,2-tetrachloroethene
Molecular Weight (g/mol) 165.82
PubChem CID 31373
Vapor Density 5.83 (vs air)
ChEBI CHEBI:17300
Formula Weight 165.83g/mol
Vapor Pressure 19mmHg (25°C), 13mmHg (20°C)
Percent Purity ≥99.8%
Chemical Name or Material Tetrachloroethylene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.