missing translation for 'onlineSavingsMsg'
Learn More

Tetrachloroethylene, ≥99.5%, Honeywell™

Product Code. p-7120865 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
4 x 2.5 L
6 x 1 L
Packaging:
Glass Bottle
Unit Size:
Pack of 4
Pack of 6
This item is not returnable. View return policy

Product Code. 15651550

Brand: Honeywell Research Chemicals 162116X1L

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

≥99.5%

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-18-4
Molecular Formula C2Cl4
Molecular Weight (g/mol) 165.82
MDL Number MFCD00000834
InChI Key CYTYCFOTNPOANT-UHFFFAOYSA-N
Synonym tetrachloroethylene, tetrachloroethene, perchloroethylene, perchlorethylene, perc, ethene, tetrachloro, tetrachlorethylene, ethylene tetrachloride, carbon dichloride, ankilostin
PubChem CID 31373
ChEBI CHEBI:17300
IUPAC Name tetrachloroethene
SMILES ClC(Cl)=C(Cl)Cl

Specifications

Melting Point -22°C
Color Colorless
Density 1.623g/mL at 25°C
Boiling Point 121°C
Packaging Glass Bottle
Linear Formula CCl2=CCl2
Refractive Index n20/D 1.505(lit.)
Quantity 6 x 1 L
UN Number UN1897
Beilstein 1361721
Vapor Density 5.83 (vs air)
Formula Weight 165.83g/mol
Vapor Pressure 13mmHg (20°C), 19mmHg (25°C)
Percent Purity ≥99.5%
Physical Form Liquid
Chemical Name or Material Tetrachloroethylene
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.