missing translation for 'onlineSavingsMsg'
Learn More

Tetrachlorocyclopropene, TRC

Product Code. 30426583
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
Unit Size:
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30426583 250 mg -
30425003 1 g 1g
30415100 500 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30426583 Supplier Toronto Research Chemicals Supplier No. TRCT291508250MG
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6262-42-6
Molecular Formula C3Cl4
Molecular Weight (g/mol) 177.84
Synonym Perchlorocyclopropene, 1,2,3,3-Tetrachlorocyclopropene
IUPAC Name 1,2,3,3-tetrachlorocyclopropene
SMILES ClC1=C(Cl)C1(Cl)Cl

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C3Cl4/c4-1-2(5)3(1,6)7
Formula Weight 175.8754
Chemical Name or Material Tetrachlorocyclopropene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.