missing translation for 'onlineSavingsMsg'
Learn More

Tetrachlorocyclopropene, 98%

Product Code. 10423361 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10423361 25 g Glass bottle 25g
10423171 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10423361 Supplier Thermo Scientific Acros Supplier No. 420190250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6262-42-6 | C3Cl4 | 177.84 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6262-42-6
Molecular Formula C3Cl4
Molecular Weight (g/mol) 177.84
MDL Number MFCD00001267
InChI Key BLZOHTXDDOAASQ-UHFFFAOYSA-N
Synonym tetrachlorocyclopropene, cyclopropene, tetrachloro, 1,2,3,3-tetrachloro-cyclopropene, tetrachlorocycloprop-1-ene, acmc-20ajjz, dsstox_cid_1660, dsstox_rid_76272, dsstox_gsid_21660, cyclopropene,1,2,3,3-tetrachloro
PubChem CID 80428
IUPAC Name 1,2,3,3-tetrachlorocyclopropene
SMILES C1(=C(C1(Cl)Cl)Cl)Cl

Specifications

Color Colorless
Boiling Point 125°C to 130°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5063 to 1.5083
Quantity 25 g
Formula Weight 177.84
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Tetrachlorocyclopropene, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Tetrachlorocyclopropene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.