missing translation for 'onlineSavingsMsg'
Learn More

Tetrachlorobis(tetrahydrofuran)titanium(IV), 98%, Thermo Scientific Chemicals

Product Code. 11324517
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11324517 2 g 2g
11304517 10 g 10g
11314517 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11324517 Supplier Thermo Scientific Alfa Aesar Supplier No. 039594.04

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 31011-57-1 | Molecular Formula: C8H16Cl4O2Ti | Molecular Weight: 333.881 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31011-57-1
Molecular Formula C8H16Cl4O2Ti
Molecular Weight (g/mol) 333.881
MDL Number MFCD00077884
InChI Key LXWBMENBONGPSB-UHFFFAOYSA-J
Synonym tetrachlorobis tetrahydrofuran titanium, tetrachlorobis tetrahydrofuran titanium iv, bis tetrahydrofuran ; titanium tetrachloride, titanium iv chloride tetrahydrofuran complex
PubChem CID 10958563
IUPAC Name oxolane;tetrachlorotitanium
SMILES C1CCOC1.C1CCOC1.Cl[Ti](Cl)(Cl)Cl

Specifications

Melting Point 126°C to 128°C
Color Yellow
Flash Point −17° (1°F)
Quantity 2 g
UN Number UN2925
Sensitivity Moisture Sensitive
Formula Weight 333.91
Percent Purity 98%
Physical Form Crystalline or Powder
Chemical Name or Material Tetrachlorobis(tetrahydrofuran)titanium(IV)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.