missing translation for 'onlineSavingsMsg'
Learn More

Tetrachloroauric(III) acid trihydrate, ACS reagent

Product Code. 10669273 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 10669273

Brand: Thermo Scientific Acros 411070050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Tetrachloroauric(III) Acid Trihydrate, >-49.0% (Au), HAuCl4.3H2O, CAS Number-16961-25-4, Gold trichloride Acid Trihydrate, Chloroauric Acid Trihydrate, 1g, Orange, 393.83, ACS Reagent, GHS Signal Word: Danger | CAS: 16961-25-4 | AuCl4H | 339.77 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16961-25-4
Molecular Formula AuCl4H
Molecular Weight (g/mol) 339.77
InChI Key VDLSFRRYNGEBEJ-UHFFFAOYSA-K
IUPAC Name trichlorogold hydrochloride
SMILES Cl.Cl[Au](Cl)Cl

Specifications

Color Orange
Physical Form Glistening Crystalline Powder or Lumps
Quantity 5 g
Linear Formula HAuCl4·3H2O
Merck Index 15,212
Synonym Chloroauric acid trihydrate, Gold trichloride acid trihydrate
Solubility Information Solubility in water: 150g/L (20°C). Other solubilities: soluble in alcohol and ether
Formula Weight 393.83
Percent Purity ≥49.0% (Au)
Grade ACS Reagent
Packaging Glass bottle
Chemical Name or Material Tetrachloroauric(III) acid trihydrate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Hydrogen tetrachloroaurate(III) trihydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.