missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetracarbonyldi-^m-chlorodirhodium(I), Rh 50.1-52.9%
Click to view available options
Quantity:
0.25 g
1 g
Unit Size:
0.25g
1g
Description
It is used widely as a chemical for research purposes as well as it acts as a a biochemical for proteomics research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Tekniske data
| Melting Point | 121°C |
| Quantity | 0.25 g |
| Sensitivity | Air and moisture sensitive |
| Solubility Information | It is soluble in organic or non-aqueous solvent. |
| Formula Weight | 388.76 |
| Physical Form | Crystalline |
| Chemical Name or Material | Tetracarbonyldi-mu-chlorodirhodium(I), Rh 50.1-52.9% |
RUO – Research Use Only
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion