missing translation for 'onlineSavingsMsg'
Learn More

Tetracarbonyldi-^m-chlorodirhodium(I), Rh 50.1-52.9%

Product Code. 11356058
Click to view available options
Quantity:
0.25 g
1 g
Unit Size:
0.25g
1g
This item is not returnable. View return policy

Product Code. 11356058

Brand: Thermo Scientific Alfa Aesar 010547.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14523-22-9 | C4Cl2O4Rh2 | 388.75 g/mol

It is used widely as a chemical for research purposes as well as it acts as a a biochemical for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14523-22-9
Molecular Formula C4Cl2O4Rh2
Molecular Weight (g/mol) 388.75
MDL Number MFCD00135610
InChI Key FGKDXBICTVUSPK-UHFFFAOYSA-L
Synonym tetracarbonyldi-mu-chlorodirhodium i, rhodium carbonyl chloride i, rhodium i carbonyl chloride, rhodium i dicarbonyl chloride dimer, di-mu-chloro-tetracarbonyldirhodium i, chlorodicarbonylrhodium i dimer, 4co.cl2rh2, bis chlorodicarbonyl rhodium, tetracarbonyldi-, i-chlorodirhodium i, tetracarbonyldi-micron-chlorodirhodium i
IUPAC Name bis(λ¹-rhodium(1+)) tetrakis(methanidylidyneoxidanium) dichloride
SMILES [Cl-].[Cl-].[Rh+].[Rh+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+]

Specifications

Melting Point 121°C
Quantity 1 g
Sensitivity Air and moisture sensitive
Solubility Information It is soluble in organic or non-aqueous solvent.
Formula Weight 388.76
Physical Form Crystalline
Chemical Name or Material Tetracarbonyldi-mu-chlorodirhodium(I), Rh 50.1-52.9%

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.