missing translation for 'onlineSavingsMsg'
Learn More

Tetracaine, 98%

Product Code. 15771759 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 15771759

Brand: Thermo Scientific Acros 460821000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 94-24-6 | C15H24N2O2 | 264.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C15H24N2O2
Molecular Weight (g/mol) 264.36
InChI Key GKCBAIGFKIBETG-UHFFFAOYSA-N
PubChem CID 5411
ChEBI CHEBI:9468

Specifications

Melting Point 43°C
Quantity 100 g
Solubility Information Solubility in water: .. Other solubilities: soluble in methanol
Formula Weight 264.36
Percent Purity 98%
Chemical Name or Material Tetracaine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.