missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylphosphonium Hydroxide (40% in Water), TRC

Product Code. 30417179
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30417179 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30417179 Supplier Toronto Research Chemicals Supplier No. TRCT28256010G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tetrabutylphosphonium Hydroxide
CAS 14518-69-5
Molecular Formula C16 H36 P . H O
Quantity 10 g
Synonym Phosphonium','tetrabutyl-','hydroxide (1:1), Phosphonium','tetrabutyl-','hydroxide (8CI,9CI), Tetrabutylphosphonium hydroxide (6CI,7CI), Tetra-n-butylphosphonium hydroxide
InChI Formula InChI=1S/C16H36P.H2O/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;/h5-16H2,1-4H3;1H2/q+1;/p-1
SMILES [OH-].CCCC[P+](CCCC)(CCCC)CCCC
IUPAC Name tetrabutylphosphanium;hydroxide
Molecular Weight (g/mol) 276.438
Formula Weight 276.258
Concentration or Composition (by Analyte or Components) 40 %
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.