missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium Tribromide, TRC

Product Code. 30414633
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30414633 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30414633 Supplier Toronto Research Chemicals Supplier No. TRCT29126050G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tetrabutylammonium Tribromide
CAS 38932-80-8
Molecular Formula C16H36Br3N
Quantity 50 g
Synonym N,N,N-Tributyl-1-butanaminium Bromide, N,N,N,N-Tetrabutylammonium Tribromide
InChI Formula InChI=1S/C16H36N.Br3/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;1-3-2/h5-16H2,1-4H3;/q+1;-1
SMILES CCCC[N+](CCCC)(CCCC)CCCC.Br[Br-]Br
Molecular Weight (g/mol) 482.18
Formula Weight 479.0398
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.