missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrabutylammonium Hydroxide, 1M in Methanol, BAKER ANALYZED™ Reagent, J.T. Baker™
Shop All J.T.Baker ProductsDescription
- For Titrant Use
Specifications
Specifications
| CAS | 2052-49-5 |
| Flash Point | 11.00°C |
| Packaging | Narrow Mouth Amber Glass Bottle |
| Molecular Formula | C16H37NO |
| MDL Number | MFCD00009425 |
| Quantity | 100 mL |
| Specific Gravity | 1L = 0.83kg |
| Synonym | tetrabutylammonium hydroxide, tetra-n-butylammonium hydroxide, tetrabutylazanium hydroxide, tetrabutylammoniumhydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide, ammonium, tetrabutyl-, hydroxide, n,n,n-tributyl-1-butanaminium hydroxide, tetra n-butyl ammonium hydroxide, tetra-n-butyl ammonium hydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide 1:1 |
| InChI Key | VDZOOKBUILJEDG-UHFFFAOYSA-M |
| SMILES | [OH-].CCCC[N+](CCCC)(CCCC)CCCC |
| Show More |
Safety and Handling
Signal Word
- Danger
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity after single exposure Category 1
- H225-Highly flammable liquid and vapour.
- H311-Toxic in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H331-Toxic if inhaled.
- H370-Causes damage to organs.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?