missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium Hydrogensulfate, TRC

Product Code. 30411289
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
Product Code. Quantity unitSize
30411289 500 g 500g
1 options
This item is not returnable. View return policy

Product Code. 30411289

Brand: Toronto Research Chemicals TRCT289220500G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tetrabutylammonium Hydrogensulfate
CAS 32503-27-8
Purity Grade Notes HPLC
Molecular Formula C16 H36 N . H O4 S
Quantity 500 g
Synonym N,N,N-Tributyl-1-butanaminium Sulfate, Tetrabutyl-ammonium Sulfate, Tetrabutylammonium Hydrogen Sulfate, TBAHS-MB, Tetra-n-butylammonium Hydrogen Sulfate, Tetrabutylammonium Bisulfate, Tetrabutylammonium Hydrosulfate
InChI Formula InChI=1S/C16H36N.H2O4S/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;1-5(2,3)4/h5-16H2,1-4H3;(H2,1,2,3,4)/q+1;/p-1
SMILES CCCC[N+](CCCC)(CCCC)CCCC.OS(=O)(=O)[O-]
IUPAC Name hydrogen sulfate;tetrabutylazanium
Molecular Weight (g/mol) 339.5343
Formula Weight 339.2443
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.