missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrabutylammonium Hydrogensulfate, TRC
Specifications
Specifications
| Chemical Name or Material | Tetrabutylammonium Hydrogensulfate |
| CAS | 32503-27-8 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C16 H36 N . H O4 S |
| Quantity | 5 g |
| Synonym | N,N,N-Tributyl-1-butanaminium Sulfate, Tetrabutyl-ammonium Sulfate, Tetrabutylammonium Hydrogen Sulfate, TBAHS-MB, Tetra-n-butylammonium Hydrogen Sulfate, Tetrabutylammonium Bisulfate, Tetrabutylammonium Hydrosulfate |
| InChI Formula | InChI=1S/C16H36N.H2O4S/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;1-5(2,3)4/h5-16H2,1-4H3;(H2,1,2,3,4)/q+1;/p-1 |
| SMILES | CCCC[N+](CCCC)(CCCC)CCCC.OS(=O)(=O)[O-] |
| IUPAC Name | hydrogen sulfate;tetrabutylazanium |
| Molecular Weight (g/mol) | 339.5343 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?