missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium hydrogen sulfate, 99%, for HPLC

Product Code. 10371351 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 g
50 g
500 g
Packaging:
Glass Bottle
Unit Size:
10g
500g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10371351 500 g Glass Bottle 500g
10095650 10 g Glass Bottle 10g
10588132 50 g Glass Bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10371351 Supplier Thermo Scientific Supplier No. 420105000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Tetrabutylammonium hydrogen sulfate, 99%, for HPLC, Packaging: Glass Bottle, Colorless to White, Grade: HPLC, Melting Point: 168 deg.C to 172 deg.C, Molecular Weight: 339.53, Percent Purity: 99% | CAS: 32503-27-8 | C16H37NO4S | 339.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ion pair chromatography, Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32503-27-8
Molecular Formula C16H37NO4S
Molecular Weight (g/mol) 339.54
MDL Number MFCD00011637
InChI Key SHFJWMWCIHQNCP-UHFFFAOYSA-M
Synonym tetrabutylammonium hydrogen sulfate, tetrabutylammonium hydrogensulfate, tetrabutylammonium bisulfate, tetra-n-butylammonium hydrogen sulfate, tbahs, tetrabutylammonium sulfate 1:1, 1-butanaminium, n,n,n-tributyl-, sulfate 1:1, tetrabutylammonium hydrogen sulphate, tetrabutylazanium hydrogen sulfate, ipc-tba-hs
PubChem CID 94433
IUPAC Name hydrogen sulfate;tetrabutylazanium
SMILES OS([O-])(=O)=O.CCCC[N+](CCCC)(CCCC)CCCC

Specifications

Absorbance (0.005M in water) (1 cm cell vs water), (0.005M in water) (1 cm cell vs water) at 0nm, 0.02 max. at 260nm, 0.03 max. at 230nm, 0.05 max. at 210nm, 0.05 max. at 220nm, 0.15 max. at 200nm
Melting Point 168°C to 172°C
Color Colorless to White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Linear Formula [CH3(CH2)3]4NHSO4
Quantity 500 g
Fieser 06,565; 07,354; 13,94
Solubility Information Solubility in water: >50%
Formula Weight 339.53
Percent Purity 99%
Grade HPLC
Physical Form Crystals or Powder
Chemical Name or Material Tetrabutylammonium hydrogen sulfate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.