missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Atrazine ELISA Kit (Colorimetric)

Product Code. 18614341 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18614341 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18614341 Supplier R&D Systems™ Supplier No. KA1414
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems.

TRUSTED_SUSTAINABILITY

Specifications

Assay Sensitivity The actual detection limit is based on the setup of experiment protocol, such as extraction procedure and dilution factor.
Product Line Novus
Product Type ELISA Kit (Colorimetric)
Sample Type Water
For Use With (Application) ELISA, Quantification
Quantity 1 Kit
Storage Requirements Store at 4°C.
Synonym 2-chloro-4-ethylamino-6-isopropylamino-1,3,5-triazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.